siRNA Details
| Gene Symbol: KRT18P55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028334.1 | 3 | 1026 | |||
| GenBank mRNA | AK097823.1 | - | 3 | 1027 | ||
| BC112026.1 | - | 3 | 647 | 8327494 | ||
| BC112028.1 | - | 3 | 635 | 8327496 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRT18P55| Gene Symbol: KRT18P55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028334.1 | 3 | 1026 | |||
| GenBank mRNA | AK097823.1 | - | 3 | 1027 | ||
| BC112026.1 | - | 3 | 647 | 8327494 | ||
| BC112028.1 | - | 3 | 635 | 8327496 | ||