siRNA Details
| Gene Symbol: SLC13A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143838.2 | NP_001137310.1 | 5, 6 | 797 | ||
| NM_001284509.1 | NP_001271438.1 | 5, 6 | 746 | |||
| NM_001284510.1 | NP_001271439.1 | 4, 5 | 668 | |||
| NM_177550.4 | NP_808218.1 | 5, 6 | 797 | |||
| XM_011523795.2 | XP_011522097.1 | 5, 6 | 787 | |||
| GenBank mRNA | AJ489980.1 | - | 4, 5 | 580 | ||
| AK127797.1 | - | 5, 6 | 694 | |||
| AK131206.1 | - | 2, 3 | 1841 | |||
| AK172785.1 | - | 5, 6 | 742 | |||
| AK297612.1 | - | 4, 5 | 613 | |||
| AY151833.1 | - | 5, 6 | 721 | |||
| BC104795.1 | - | 5, 6 | 753 | 8143798 | ||
| BC112151.1 | - | 5, 6 | 753 | 8327619 | ||
| BC143689.1 | - | 5, 6 | 753 | 9052203 | ||

SLC13A5
Genomic Map