siRNA Details
| Gene Symbol: TMEM102 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320444.1 | NP_001307373.1 | 2 | 808 | ||
| NM_178518.2 | NP_848613.1 | 3 | 682 | |||
| GenBank mRNA | AK094197.1 | - | 2 | 531 | ||
| BC046189.1 | - | 3 | 682 | 5730428 | ||

TMEM102
Genomic Map