siRNA Details
| Gene Symbol: PTRF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012232.5 | NP_036364.2 | 1 | 676 | ||
| XM_005257242.3 | XP_005257299.1 | 1 | 676 | |||
| GenBank mRNA | AF312393.1 | - | 1 | 873 | ||
| AK000715.1 | - | 1 | 162 | |||
| AK092406.1 | - | 2 | 477 | |||
| AK298072.1 | - | 2 | 557 | |||
| AK298563.1 | - | 2 | 531 | |||
| AK314389.1 | - | 1 | 612 | |||
| BC066123.1 | - | 1 | 612 | 5269172 | ||
| BC073759.1 | - | 203 | 6572478 | |||

PTRF
Genomic Map