siRNA Details
Gene Symbol: TMEM105 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178520.3 | NP_848615.1 | 3 | 638 | ||
GenBank mRNA | AK096111.1 | - | 3 | 638 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TMEM105 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178520.3 | NP_848615.1 | 3 | 638 | ||
GenBank mRNA | AK096111.1 | - | 3 | 638 |