siRNA Details
| Gene Symbol: PRR19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199285.2 | NP_954979.2 | 3 | 1175 | ||
| XM_005258779.4 | XP_005258836.2 | 3 | 1113 | |||
| XM_005258780.3 | XP_005258837.1 | 3 | 910 | |||
| XM_006723154.3 | XP_006723217.1 | 3 | 1058 | |||
| XM_011526787.1 | XP_011525089.1 | 3 | 876 | |||
| XM_011526788.1 | XP_011525090.1 | 3 | 887 | |||
| XM_011526789.2 | XP_011525091.1 | 4 | 879 | |||
| XM_011526790.2 | XP_011525092.1 | 3 | 898 | |||
| GenBank mRNA | AI208291.1 | - | 1 | 354 | ||
| AK124116.1 | - | 2 | 1608 | |||
| AK291528.1 | - | 2 | 1261 | |||
| BC063021.1 | - | 2 | 1294 | 5245434 | ||

PRR19
Genomic Map