siRNA Details
| Gene Symbol: TMEM145 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173633.2 | NP_775904.2 | 7 | 616 | ||
| XM_005258781.4 | XP_005258838.1 | 7 | 658 | |||
| XM_011526791.2 | XP_011525093.1 | 7 | 610 | |||
| XM_011526792.1 | XP_011525094.1 | 7 | 658 | |||
| GenBank mRNA | AK075286.1 | - | 7 | 605 | ||

TMEM145
Genomic Map