siRNA Details
Gene Symbol: CXCL17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198477.2 | NP_940879.1 | 2, 3 | 365 | ||
GenBank mRNA | AK292985.1 | - | 2, 3 | 263 | ||
AY358433.1 | - | 2, 3 | 301 | |||
AY598464.1 | - | 2, 3 | 365 | |||
BC093946.1 | - | 2, 3 | 216 | 7939791 | ||
BC112095.1 | - | 2, 3 | 218 | 8327563 |
