siRNA Details
| Gene Symbol: WDR62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083961.1 | NP_001077430.1 | 6 | 714 | ||
| NM_173636.4 | NP_775907.4 | 6 | 714 | |||
| XM_005258809.2 | XP_005258866.1 | 6 | 714 | |||
| XM_011526837.1 | XP_011525139.1 | 6 | 714 | |||
| XM_011526838.1 | XP_011525140.1 | 6 | 714 | |||
| XM_011526839.1 | XP_011525141.1 | 6 | 714 | |||
| XM_017026665.1 | XP_016882154.1 | 6 | 714 | |||
| GenBank mRNA | AK090617.1 | - | 6 | 714 | ||
| BC058939.1 | - | 6 | 658 | |||
| BU508347.1 | - | 6 | 477 | |||
| BX647726.1 | - | 6 | 633 | |||

WDR62
Genomic Map