siRNA Details
| Gene Symbol: ZFP82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321917.1 | NP_001308846.1 | 5 | 720 | ||
| NM_133466.3 | NP_597723.1 | 5 | 717 | |||
| GenBank mRNA | AB075828.1 | - | 1 | 2184 | ||
| AK074942.1 | - | 5 | 699 | |||
| AK315709.1 | - | 5 | 677 | |||
| AL834267.1 | - | 5 | 691 | |||
| BC032577.2 | - | 5 | 684 | 5492906 | ||
| GU388429.1 | - | 5 | 687 | |||

ZFP82
Genomic Map