siRNA Details
| Gene Symbol: NWD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007525.3 | NP_001007526.3 | 6 | 983 | ||
| NM_001290355.1 | NP_001277284.1 | 7 | 1635 | |||
| XM_011527929.1 | XP_011526231.1 | 7 | 1167 | |||
| XM_011527930.2 | XP_011526232.1 | 7 | 1653 | |||
| XM_011527931.2 | XP_011526233.1 | 6 | 1433 | |||
| XM_011527935.1 | XP_011526237.1 | 7 | 1167 | |||
| XM_017026669.1 | XP_016882158.1 | 8 | 1720 | |||
| GenBank mRNA | BC117698.1 | - | 5 | 631 | 40073616 | |
| BX648940.1 | - | 8 | 1635 | |||
| CR749794.1 | - | 6 | 979 | |||

NWD1
Genomic Map