siRNA Details
| Gene Symbol: RSPO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038633.3 | NP_001033722.1 | 5 | 1044 | ||
| NM_001242908.1 | NP_001229837.1 | 4 | 865 | |||
| NM_001242909.1 | NP_001229838.1 | 4 | 544 | |||
| NM_001242910.1 | NP_001229839.1 | 4 | 865 | |||
| XM_006710583.3 | XP_006710646.1 | 3 | 780 | |||
| GenBank mRNA | AK098225.1 | - | 4 | 286 | ||
| BC114966.1 | - | 3 | 462 | 40084277 | ||
| DQ165084.1 | - | 4 | 607 | |||
| DQ165085.1 | - | 4 | 607 | |||
| DQ318235.1 | - | 5 | 754 | |||

RSPO1
Genomic Map