siRNA Details
| Gene Symbol: C1orf111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182581.3 | NP_872387.2 | 1, 2 | 173 | ||
| XM_011509443.2 | XP_011507745.1 | 1, 2 | 281 | |||
| XM_011509445.2 | XP_011507747.1 | 1, 2 | 281 | |||
| GenBank mRNA | AY248900.1 | - | 1, 2 | 152 | ||
| BC032957.1 | - | 1, 2 | 180 | 4827863 | ||

C1orf111
Genomic Map