siRNA Details
| Gene Symbol: BTBD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183242.3 | NP_899065.2 | 8 | 1255 | ||
| GenBank mRNA | AK296516.1 | - | 7 | 925 | ||
| BC013922.1 | - | 8 | 1228 | 4048340 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BTBD8| Gene Symbol: BTBD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183242.3 | NP_899065.2 | 8 | 1255 | ||
| GenBank mRNA | AK296516.1 | - | 7 | 925 | ||
| BC013922.1 | - | 8 | 1228 | 4048340 | ||