siRNA Details
| Gene Symbol: ZNF663P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045983.1 | 1 | 824 | |||
| GenBank mRNA | AL831934.1 | - | 3 | 1060 | ||
| BC100775.1 | - | 1 | 824 | |||
| BC100776.2 | - | 1 | 824 | |||
| BC100777.1 | - | 1 | 824 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF663P| Gene Symbol: ZNF663P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045983.1 | 1 | 824 | |||
| GenBank mRNA | AL831934.1 | - | 3 | 1060 | ||
| BC100775.1 | - | 1 | 824 | |||
| BC100776.2 | - | 1 | 824 | |||
| BC100777.1 | - | 1 | 824 | |||