siRNA Details
| Gene Symbol: C20orf203 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182584.2 | NP_872390.2 | 3 | 411 | ||
| GenBank mRNA | AK091025.1 | - | 3 | 308 | ||
| BC105014.1 | - | 3 | 187 | |||
| BC105042.1 | - | 3 | 189 | |||
| GU931819.1 | - | 4 | 721 | |||
| GU931820.1 | - | 4 | 721 | |||

C20orf203
Genomic Map