siRNA Details
| Gene Symbol: KRTAP13-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181600.1 | NP_853631.1 | 1 | 450 | ||
| GenBank mRNA | AB096941.1 | - | 1 | 472 | ||
| BC131691.1 | - | 1 | 454 | 40119689 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRTAP13-4| Gene Symbol: KRTAP13-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181600.1 | NP_853631.1 | 1 | 450 | ||
| GenBank mRNA | AB096941.1 | - | 1 | 472 | ||
| BC131691.1 | - | 1 | 454 | 40119689 | ||