siRNA Details
| Gene Symbol: AATBC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026961.1 | 1 | 982 | |||
| GenBank mRNA | AK128598.1 | - | 1 | 983 | ||
| BC111558.1 | - | 1 | 981 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AATBC| Gene Symbol: AATBC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026961.1 | 1 | 982 | |||
| GenBank mRNA | AK128598.1 | - | 1 | 983 | ||
| BC111558.1 | - | 1 | 981 | |||