siRNA Details
| Gene Symbol: C2orf61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163561.1 | NP_001157033.1 | 4 | 557 | ||
| NM_173649.2 | NP_775920.1 | 4 | 557 | |||
| GenBank mRNA | AK097491.1 | - | 4 | 466 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C2orf61| Gene Symbol: C2orf61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163561.1 | NP_001157033.1 | 4 | 557 | ||
| NM_173649.2 | NP_775920.1 | 4 | 557 | |||
| GenBank mRNA | AK097491.1 | - | 4 | 466 | ||