siRNA Details
| Gene Symbol: C3orf38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173824.3 | NP_776185.2 | 2 | 474 | ||
| XM_005264745.4 | XP_005264802.1 | 2 | 503 | |||
| GenBank mRNA | AK297719.1 | - | 2 | 230 | ||
| AK313547.1 | - | 2 | 215 | |||
| AL832398.1 | - | 2 | 474 | |||
| BC024188.2 | - | 2 | 258 | 4824439 | ||
| KU178900.1 | - | 2 | 157 | |||
| KU178901.1 | - | 2 | 157 | |||

C3orf38
Genomic Map