siRNA Details
| Gene Symbol: RFT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052859.3 | NP_443091.1 | 13 | 3399 | ||
| GenBank mRNA | AK098811.1 | - | 1 | 591 | ||
| AL713685.1 | - | 1 | 538 | |||
| BC101247.2 | - | 1 | 464 | |||
| BC101248.2 | - | 1 | 464 | |||
| BC101249.2 | - | 1 | 464 | |||
| BC101250.1 | - | 1 | 464 | |||
| BC113058.1 | - | 1 | 464 | |||
| BC113059.1 | - | 1 | 464 | |||

RFT1
Genomic Map