siRNA Details
| Gene Symbol: DOK7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164673.1 | NP_001158145.1 | 7 | 1546 | ||
| NM_001256896.1 | NP_001243825.1 | 4 | 1255 | |||
| NM_173660.4 | NP_775931.3 | 7 | 1557 | |||
| GenBank mRNA | AB220918.1 | - | 7 | 1487 | ||
| AK091037.1 | - | 7 | 1541 | |||
| AK096456.1 | - | 4 | 1255 | |||
| BC043568.1 | - | 3 | 894 | |||
| BC062369.1 | - | 3 | 949 | |||
| BC131544.1 | - | 7 | 1494 | 40085498 | ||
| BC141852.1 | - | 7 | 1528 | 8860352 | ||

DOK7
Genomic Map