siRNA Details
| Gene Symbol: RNF175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173662.2 | NP_775933.1 | 8 | 1165 | ||
| XM_005262939.3 | XP_005262996.1 | 7 | 967 | |||
| XM_005262940.4 | XP_005262997.1 | 6 | 1104 | |||
| XM_011531879.2 | XP_011530181.1 | 8 | 1110 | |||
| XM_011531881.2 | XP_011530183.1 | 8 | 1249 | |||
| XM_011531883.2 | XP_011530185.1 | 8 | 1130 | |||
| XM_017008047.1 | XP_016863536.1 | 6 | 883 | |||
| GenBank mRNA | BC034385.1 | - | 8 | 1166 | 5180678 | |

RNF175
Genomic Map