siRNA Details
| Gene Symbol: C5orf51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175921.5 | NP_787117.3 | 6 | 696 | ||
| XM_005248289.4 | XP_005248346.1 | 5 | 612 | |||
| XM_011514032.2 | XP_011512334.1 | 7 | 825 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C5orf51| Gene Symbol: C5orf51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175921.5 | NP_787117.3 | 6 | 696 | ||
| XM_005248289.4 | XP_005248346.1 | 5 | 612 | |||
| XM_011514032.2 | XP_011512334.1 | 7 | 825 | |||