siRNA Details
| Gene Symbol: ZNF454 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001178089.2 | NP_001171560.1 | 5 | 770 | ||
| NM_001178090.2 | NP_001171561.1 | 5 | 709 | |||
| NM_001323306.1 | NP_001310235.1 | 5 | 767 | |||
| NM_182594.3 | NP_872400.2 | 5 | 712 | |||
| GenBank mRNA | AK094763.1 | - | 5 | 680 | ||
| AK292317.1 | - | 5 | 738 | |||
| BC105061.1 | - | 4 | 472 | 8144064 | ||
| BC112276.1 | - | 4 | 474 | 8327744 | ||

ZNF454
Genomic Map