siRNA Details
| Gene Symbol: LOC285696 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027253.1 | 1 | 973 | |||
| GenBank mRNA | AK091366.1 | - | 1 | 598 | ||
| AK125192.1 | - | 1 | 952 | |||
| BC093861.1 | - | 1 | 572 | |||
| BC093887.1 | - | 1 | 572 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC285696| Gene Symbol: LOC285696 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027253.1 | 1 | 973 | |||
| GenBank mRNA | AK091366.1 | - | 1 | 598 | ||
| AK125192.1 | - | 1 | 952 | |||
| BC093861.1 | - | 1 | 572 | |||
| BC093887.1 | - | 1 | 572 | |||