siRNA Details
| Gene Symbol: CAGE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170692.1 | NP_001164163.1 | 4 | 895 | ||
| NM_001170693.1 | NP_001164164.1 | 4 | 895 | |||
| NM_205864.2 | NP_995586.1 | 3 | 677 | |||
| GenBank mRNA | AF414185.1 | - | 3 | 676 | ||
| AK097760.1 | - | 4 | 594 | |||
| BC026188.1 | - | 4 | 573 | |||
| BC026194.2 | - | 4 | 580 | 4826784 | ||
| BC033545.1 | - | 4 | 580 | 4826548 | ||

CAGE1
Genomic Map