siRNA Details
| Gene Symbol: SCARA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173833.5 | NP_776194.2 | 2 | 668 | ||
| GenBank mRNA | AK172746.1 | - | 2 | 498 | ||
| AY337579.1 | - | 1 | 57 | |||
| AY358150.1 | - | 2 | 491 | |||
| BC033153.1 | - | 2 | 440 | 4138639 | ||

SCARA5
Genomic Map