siRNA Details
| Gene Symbol: SCAI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144877.2 | NP_001138349.1 | 18 | 1920 | ||
| NM_173690.4 | NP_775961.2 | 19 | 1989 | |||
| GenBank mRNA | AF086497.1 | - | 5 | 534 | ||
| AK093983.1 | - | 18 | 1879 | |||
| AK098024.1 | - | 19 | 1926 | |||
| BC104030.2 | - | 19 | 1917 | 40025626 | ||
| BC104031.1 | - | 18 | 1848 | 40025627 | ||
| BC104032.1 | - | 18 | 1766 | 40025629 | ||

SCAI
Genomic Map