siRNA Details
| Gene Symbol: FAM78A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033387.3 | NP_203745.2 | 1 | 458 | ||
| GenBank mRNA | AK095423.1 | - | 1 | 486 | ||
| BC049388.2 | - | 1 | 458 | 6050655 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM78A| Gene Symbol: FAM78A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033387.3 | NP_203745.2 | 1 | 458 | ||
| GenBank mRNA | AK095423.1 | - | 1 | 486 | ||
| BC049388.2 | - | 1 | 458 | 6050655 | ||