siRNA Details
| Gene Symbol: TRIM59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173084.2 | NP_775107.1 | 3 | 1304 | ||
| GenBank mRNA | AK291284.1 | - | 3 | 1226 | ||
| AY159379.1 | - | 3 | 1304 | |||
| BC109259.1 | - | 3 | 1291 | 40025031 | ||
| BC109260.1 | - | 3 | 1291 | 40025036 | ||

TRIM59
Genomic Map