siRNA Details
Gene Symbol: VN1R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173856.2 | NP_776255.2 | 1 | 446 | ||
GenBank mRNA | BC130356.1 | - | 1 | 446 | 40146387 | |
BC130358.1 | - | 1 | 446 | 40146389 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: VN1R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173856.2 | NP_776255.2 | 1 | 446 | ||
GenBank mRNA | BC130356.1 | - | 1 | 446 | 40146387 | |
BC130358.1 | - | 1 | 446 | 40146389 |