siRNA Details
| Gene Symbol: VN1R5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173858.1 | NP_776257.1 | 1 | 167 | ||
| GenBank mRNA | BC126341.1 | - | 1 | 267 | 8992057 | |
| BC126367.1 | - | 1 | 267 | 8992083 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

VN1R5| Gene Symbol: VN1R5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173858.1 | NP_776257.1 | 1 | 167 | ||
| GenBank mRNA | BC126341.1 | - | 1 | 267 | 8992057 | |
| BC126367.1 | - | 1 | 267 | 8992083 | ||