siRNA Details
| Gene Symbol: TAAR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175067.1 | NP_778237.1 | 1 | 165 | ||
| GenBank mRNA | AY183470.1 | - | 1 | 165 | ||
| BC069157.1 | - | 1 | 165 | 7217018 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TAAR6| Gene Symbol: TAAR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175067.1 | NP_778237.1 | 1 | 165 | ||
| GenBank mRNA | AY183470.1 | - | 1 | 165 | ||
| BC069157.1 | - | 1 | 165 | 7217018 | ||
Set Membership: |
|