siRNA Details
| Gene Symbol: PPM1J | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005167.5 | NP_005158.5 | 4 | 821 | ||
| GenBank mRNA | AB451327.1 | - | 3 | 176 | ||
| AB451471.1 | - | 3 | 176 | |||
| AK093270.1 | - | 4 | 801 | |||
| AL133644.1 | - | 5 | 1585 | |||
| BC011803.2 | - | 4 | 745 | |||
| BC064628.1 | - | 4 | 751 | |||
| BC073828.1 | - | 4 | 813 | 5244363 | ||

PPM1J
Genomic Map