siRNA Details
| Gene Symbol: CLEC4D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080387.4 | NP_525126.2 | 6 | 778 | ||
| XM_011520632.2 | XP_011518934.1 | 7 | 747 | |||
| GenBank mRNA | AF411850.1 | - | 6 | 648 | ||
| AK314071.1 | - | 6 | 764 | |||
| AY115592.1 | - | 6 | 769 | |||
| AY297446.1 | - | 6 | 777 | |||
| AY486482.1 | - | 6 | 602 | |||
| AY486483.1 | - | 6 | 602 | |||
| BC032313.1 | - | 6 | 778 | 5218660 | ||

CLEC4D
Genomic Map