siRNA Details
| Gene Symbol: RAB7B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164522.2 | NP_001157994.1 | 3 | 367 | ||
| NM_001304839.1 | NP_001291768.1 | 3 | 367 | |||
| NM_177403.5 | NP_796377.3 | 2 | 441 | |||
| XM_006711288.3 | XP_006711351.1 | 3 | 364 | |||
| GenBank mRNA | AK122657.1 | - | 3 | 362 | ||
| AK127140.1 | - | 2 | 425 | |||
| AY094596.1 | - | 3 | 314 | |||
| BC017092.1 | - | 3 | 309 | 3851998 | ||
| BI757251.1 | - | 2 | 438 | |||
| DA966224.1 | - | 3 | 367 | |||

RAB7B
Genomic Map