siRNA Details
| Gene Symbol: TAS2R60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177437.1 | NP_803186.1 | 1 | 299 | ||
| GenBank mRNA | BC100938.2 | - | 1 | 299 | 40004056 | |
| BC100939.2 | - | 1 | 299 | 40004057 | ||
| BC100940.1 | - | 1 | 299 | 40004058 | ||

TAS2R60
Genomic Map