siRNA Details
| Gene Symbol: ST8SIA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004470.1 | NP_001004470.1 | 8 | 1064 | ||
| XM_017016174.1 | XP_016871663.1 | 7 | 1056 | |||
| GenBank mRNA | AB062101.1 | - | 8 | 1064 | ||
| AJ621583.1 | - | 8 | 1064 | |||
| BC137102.1 | - | 8 | 1196 | 9021099 | ||
| BC137103.1 | - | 8 | 1196 | 9021100 | ||
| BC144472.1 | - | 7 | 1083 | |||
| BX951710.1 | - | 1 | 62 | |||

ST8SIA6
Genomic Map