siRNA Details
| Gene Symbol: JMJD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005920.2 | NP_001005920.2 | 7 | 675 | ||
| NM_001323918.1 | NP_001310847.1 | 7 | 682 | |||
| NM_001323919.1 | NP_001310848.1 | 7 | 682 | |||
| NM_001323920.1 | NP_001310849.1 | 6 | 592 | |||
| NM_001323922.1 | NP_001310851.1 | 6 | 592 | |||
| NR_136650.1 | 6 | 770 | ||||
| NR_136651.1 | 6 | 770 | ||||
| NR_136652.1 | 5 | 680 | ||||
| GenBank mRNA | AF370420.1 | - | 7 | 559 | ||
| AL524336.3 | - | 6 | 655 | |||
| AL524922.3 | - | 6 | 657 | |||
| BC073785.1 | - | 6 | 647 | |||
| BC094850.1 | - | 4 | 293 | |||
| BC137100.1 | - | 7 | 641 | 9021097 | ||
| BC137101.1 | - | 7 | 641 | 9021098 | ||
| BG284477.1 | - | 2 | 132 | |||
| BG423721.1 | - | 3 | 192 | |||
| BG481039.1 | - | 7 | 578 | |||
| BG826580.1 | - | 4 | 473 | |||
| BI463867.1 | - | 7 | 682 | |||
| BI767435.1 | - | 6 | 495 | |||
| BX335015.2 | - | 7 | 561 | |||
| DQ074695.1 | - | 6 | 465 | |||

JMJD8
Genomic Map