siRNA Details
| Gene Symbol: CPLX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181654.3 | NP_857637.1 | 2, 3 | 446 | ||
| GenBank mRNA | AB593158.1 | - | 2, 3 | 427 | ||
| AK123184.1 | - | 2, 3 | 446 | |||
| AY286502.1 | - | 2, 3 | 276 | |||
| BC106886.2 | - | 1, 2 | 419 | |||
| BC106887.1 | - | 2, 3 | 439 | 40030305 | ||
| BC106888.1 | - | 2, 3 | 439 | 40030309 | ||
| BX640655.1 | - | 2, 3 | 425 | |||
| BX648518.1 | - | 2, 3 | 429 | |||

CPLX4
Genomic Map