siRNA Details
| Gene Symbol: ZNF546 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178544.4 | NP_848639.2 | 3 | 291 | ||
| XM_011526899.2 | XP_011525201.1 | 3 | 203 | |||
| GenBank mRNA | AK292528.1 | - | 3 | 239 | ||
| BC045649.1 | - | 3 | 205 | 5268795 | ||

ZNF546
Genomic Map