siRNA Details
| Gene Symbol: RXFP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181885.2 | NP_871001.1 | 1 | 180 | ||
| GenBank mRNA | AY170824.1 | - | 1 | 159 | ||
| AY288415.1 | - | 1 | 159 | |||
| AY394502.1 | - | 1 | 159 | |||
| BC101507.1 | - | 1 | 179 | 8069013 | ||
| BC101509.1 | - | 1 | 179 | 8069015 | ||
| DN693323.1 | - | 1 | 180 | |||
| EU432129.1 | - | 1 | 159 | |||
More Information
Set Membership: |
|

RXFP4
Genomic Map