siRNA Details
| Gene Symbol: ANKRD45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198493.2 | NP_940895.1 | 4 | 606 | ||
| XM_011509466.2 | XP_011507768.1 | 5 | 1053 | |||
| XM_011509467.1 | XP_011507769.1 | 6 | 1577 | |||
| XM_011509468.1 | XP_011507770.1 | 5 | 1545 | |||
| XM_011509470.1 | XP_011507772.1 | 4 | 1261 | |||
| XM_017001123.1 | XP_016856612.1 | 6 | 915 | |||
| GenBank mRNA | AK127170.1 | - | 4 | 606 | ||
| BC126353.1 | - | 4 | 606 | 8992069 | ||
| BC126355.1 | - | 4 | 606 | 8992071 | ||

ANKRD45
Genomic Map