siRNA Details
| Gene Symbol: KLHL17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198317.2 | NP_938073.1 | 2, 3 | 473 | ||
| XM_006710600.3 | XP_006710663.1 | 2, 3 | 957 | |||
| XM_006710601.3 | XP_006710664.1 | 2, 3 | 957 | |||
| GenBank mRNA | AY423763.1 | - | 2, 3 | 473 | ||

KLHL17
Genomic Map