siRNA Details
| Gene Symbol: KLHL17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198317.2 | NP_938073.1 | 4, 5 | 818 | ||
| GenBank mRNA | AK127585.1 | - | 3, 4 | 465 | ||
| AY423763.1 | - | 4, 5 | 818 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KLHL17| Gene Symbol: KLHL17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198317.2 | NP_938073.1 | 4, 5 | 818 | ||
| GenBank mRNA | AK127585.1 | - | 3, 4 | 465 | ||
| AY423763.1 | - | 4, 5 | 818 | |||