siRNA Details
| Gene Symbol: SLC35E4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001479.3 | NP_001001479.1 | 1 | 827 | ||
| NM_001318370.1 | NP_001305299.1 | 1 | 827 | |||
| NM_001318371.1 | NP_001305300.1 | 1 | 827 | |||
| XM_017028794.1 | XP_016884283.1 | 2 | 1919 | |||
| XM_017028795.1 | XP_016884284.1 | 2 | 1493 | |||
| GenBank mRNA | BC040191.1 | - | 1 | 835 | 4814106 | |
| BC093099.1 | - | 1 | 319 | 30511864 | ||
| BC107119.2 | - | 1 | 213 | 40023535 | ||
| CR456351.1 | - | 1 | 316 | |||

SLC35E4
Genomic Map