siRNA Details
| Gene Symbol: TEX33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163857.1 | NP_001157329.1 | 3 | 570 | ||
| NM_178552.3 | NP_848647.1 | 3 | 551 | |||
| XM_011530165.2 | XP_011528467.1 | 4 | 783 | |||
| XM_011530166.1 | XP_011528468.1 | 3 | 526 | |||
| GenBank mRNA | BC042635.1 | - | 3 | 513 | 5171722 | |
| CR456416.1 | - | 3 | 536 | |||

TEX33
Genomic Map