siRNA Details
| Gene Symbol: RSPO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178565.4 | NP_848660.3 | 2, 3 | 745 | ||
| XM_017013395.1 | XP_016868884.1 | 2, 3 | 734 | |||
| GenBank mRNA | AK123023.1 | - | 2, 3 | 708 | ||
| AY358166.1 | - | 2, 3 | 398 | |||

RSPO2
Genomic Map