siRNA Details
| Gene Symbol: RSPO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282863.1 | NP_001269792.1 | 4 | 829 | ||
| NM_001317942.1 | NP_001304871.1 | 4 | 551 | |||
| NM_178565.4 | NP_848660.3 | 5 | 1230 | |||
| XM_011517018.1 | XP_011515320.1 | 5 | 1246 | |||
| XM_011517019.1 | XP_011515321.1 | 5 | 1243 | |||
| XM_017013395.1 | XP_016868884.1 | 5 | 1216 | |||
| GenBank mRNA | AK123023.1 | - | 5 | 1193 | ||
| AK123027.1 | - | 3 | 360 | |||
| AY358166.1 | - | 5 | 883 | |||
| BC027938.1 | - | 4 | 551 | 5201681 | ||
| BC036554.1 | - | 4 | 830 | 5265957 | ||

RSPO2
Genomic Map